Assay Details
| Gene Symbol: Tead1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198589.1 | NP_001185518.1 | 9 - 10 | 980 | 71 | |
| XM_006230052.2 | XP_006230114.1 | 10 - 11 | 1325 | 71 | ||
| XM_006230053.2 | XP_006230115.1 | 9 - 10 | 1313 | 71 | ||
| XM_006230054.2 | XP_006230116.1 | 9 - 10 | 1262 | 71 | ||
| XM_006230055.2 | XP_006230117.1 | 9 - 10 | 1262 | 71 | ||
| XM_006230056.2 | XP_006230118.1 | 8 - 9 | 1250 | 71 | ||
| XM_006230057.2 | XP_006230119.1 | 8 - 9 | 1250 | 71 | ||
| XM_006230058.2 | XP_006230120.1 | 7 - 8 | 1187 | 71 | ||
| XM_006230059.2 | XP_006230121.1 | 10 - 11 | 2069 | 71 | ||
| XM_008759767.1 | XP_008757989.1 | 10 - 11 | 1519 | 71 | ||
| XM_008759768.1 | XP_008757990.1 | 10 - 11 | 882 | 71 | ||
| GenBank mRNA | AY529194.1 | - | 8 - 9 | 710 | 71 | |
| AY529195.1 | - | 8 - 9 | 710 | 71 | ||
| AY529196.1 | - | 7 - 8 | 698 | 71 | ||
| AY529197.1 | - | 7 - 8 | 647 | 71 | ||
| AY529198.1 | - | 6 - 7 | 635 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead1
Genomic Map