Assay Details
| Gene Symbol: Atp7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012511.2 | NP_036643.2 | 6 - 7 | 1928 | 66 | |
| XM_006253346.2 | XP_006253408.1 | 6 - 7 | 2604 | 66 | ||
| GenBank mRNA | AB017790.1 | - | 6 - 7 | 1928 | 66 | |
| AB017791.1 | - | 6 - 7 | 1928 | 66 | ||
| AB017792.1 | - | 6 - 7 | 1928 | 66 | ||
| AB017793.1 | - | 6 - 7 | 1928 | 66 | ||
| U08344.1 | - | 6 - 7 | 1944 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp7b
Genomic Map