Assay Details
| Gene Symbol: Runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278483.1 | NP_001265412.1 | 4 - 5 | 802 | 86 | |
| XM_006244549.2 | XP_006244611.1 | 3 - 4 | 1003 | 86 | ||
| XM_006244550.2 | XP_006244612.1 | 4 - 5 | 1014 | 86 | ||
| XM_006244551.2 | XP_006244613.1 | 4 - 5 | 981 | 86 | ||
| XM_006244554.2 | XP_006244616.1 | 2 - 3 | 583 | 86 | ||
| GenBank mRNA | AB025797.1 | - | 2 - 3 | 179 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Runx2
Genomic Map