Assay Details
| Gene Symbol: Itpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107774.1 | NP_001101244.1 | 6 - 7 | 475 | 59 | |
| GenBank mRNA | FQ230286.1 | - | 6 - 7 | 438 | 59 | |
| FQ231907.1 | - | 6 - 7 | 438 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itpa
Genomic Map