Assay Details
| Gene Symbol: Mettl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114181.1 | NP_001107653.1 | 2 - 3 | 444 | 85 | |
| GenBank mRNA | BC158754.1 | - | 2 - 3 | 444 | 9102963 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl5
Genomic Map