Assay Details
| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106485.3 | NP_001099955.2 | 2 - 3 | 430 | 92 | |
| GenBank mRNA | FQ229205.1 | - | 2 - 3 | 414 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp7
Genomic Map