Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077429.1 | NP_001070897.1 | 12 - 13 | 1161 | 93 | |
| XM_006234385.2 | XP_006234447.1 | 12 - 13 | 1245 | 93 | ||
| GenBank mRNA | BC126100.1 | - | 12 - 13 | 1161 | 7132654 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map