Assay Details
| Gene Symbol: Tp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107749.1 | NP_001101219.1 | 2 - 3 | 226 | 60 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107749.1 transcripts.
Set Membership: |
|

Tp53i11
Genomic Map