Assay Details
| Gene Symbol: Arhgap11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168524.1 | NP_001161996.1 | 4 - 5 | 810 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap11a| Gene Symbol: Arhgap11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168524.1 | NP_001161996.1 | 4 - 5 | 810 | 105 | |
Set Membership: |
|