Assay Details
| Gene Symbol: Lpcat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106494.1 | NP_001099964.1 | 11 - 12 | 1618 | 78 | |
| XM_006234702.2 | XP_006234764.1 | 11 - 12 | 1640 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat4
Genomic Map