Assay Details
| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024780.1 | NP_001019951.1 | 5 - 6 | 639 | 73 | |
| GenBank mRNA | BC097446.1 | - | 5 - 6 | 639 | 7456528 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Prkra
Genomic Map