Assay Details
| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024780.1 | NP_001019951.1 | 3 - 4 | 447 | 85 | |
| GenBank mRNA | BC097446.1 | - | 3 - 4 | 447 | 7456528 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Prkra
Genomic Map