Assay Details
| Gene Symbol: Kif17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750065.3 | XP_003750113.1 | 7 - 8 | 1414 | 57 | |
| XM_006239290.2 | XP_006239352.1 | 7 - 8 | 1414 | 57 | ||
| XM_006239291.2 | XP_006239353.1 | 7 - 8 | 1414 | 57 | ||
| XM_006239292.2 | XP_006239354.1 | 7 - 8 | 1414 | 57 | ||
More Information
Set Membership: |
|

Kif17
Genomic Map