Assay Details
| Gene Symbol: Slc35e2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107998.1 | NP_001101468.1 | 2 - 3 | 1225 | 138 | |
| XM_006239572.2 | XP_006239634.1 | 2 - 3 | 787 | 138 | ||
| XM_006239573.2 | XP_006239635.1 | 2 - 3 | 904 | 138 | ||
| XM_008764361.1 | XP_008762583.1 | 2 - 3 | 904 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc35e2b
Genomic Map