Assay Details
| Gene Symbol: Slc35e2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107998.1 | NP_001101468.1 | 6 - 7 | 1659 | 116 | |
| XM_006239572.2 | XP_006239634.1 | 6 - 7 | 1221 | 116 | ||
| XM_006239573.2 | XP_006239635.1 | 6 - 7 | 1338 | 116 | ||
| XM_008764361.1 | XP_008762583.1 | 6 - 7 | 1338 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc35e2b
Genomic Map