Assay Details
| Gene Symbol: Meaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113784.1 | NP_001107256.1 | 1 - 2 | 96 | 95 | |
| XM_006238886.2 | XP_006238948.1 | 1 - 2 | 356 | 95 | ||
| XM_006238887.2 | XP_006238949.1 | 1 - 2 | 356 | 95 | ||
| GenBank mRNA | BC158755.1 | - | 1 - 2 | 96 | 9103162 | 95 |
| FQ215854.1 | - | 1 - 2 | 114 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Meaf6
Genomic Map