Assay Details
| Gene Symbol: Dffa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053679.2 | NP_446131.2 | 3 - 4 | 506 | 98 | |
| XM_006239362.2 | XP_006239424.1 | 3 - 4 | 527 | 98 | ||
| GenBank mRNA | AF136601.1 | - | 3 - 4 | 491 | 98 | |
| BC100067.1 | - | 3 - 4 | 506 | 7389030 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dffa
Genomic Map