Assay Details
| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133310.2 | NP_579844.2 | 2 - 3 | 336 | 100 | |
| GenBank mRNA | CB616023.1 | - | 2 - 3 | 336 | 100 | |
More Information
Set Membership: |
|

Icmt
Genomic Map