Assay Details
| Gene Symbol: Mrpl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109428.1 | NP_001102898.1 | 1 - 2 | 216 | 71 | |
| GenBank mRNA | BC166541.1 | - | 1 - 2 | 101 | 7192827 | 71 |
| FQ224518.1 | - | 1 - 2 | 120 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl20
Genomic Map