Assay Details
| Gene Symbol: LOC299271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181630.2 | NP_853661.1 | 1 - 2 | 245 | 94 | |
| GenBank mRNA | AY113703.1 | - | 1 - 2 | 245 | 94 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY113703.1,NM_181630.2 transcripts.
Set Membership: |
|

LOC299271
Genomic Map