Assay Details
| Gene Symbol: Slirp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109507.1 | NP_001102977.1 | 1 - 2 | 195 | 81 | |
| GenBank mRNA | BC166913.1 | - | 1 - 2 | 106 | 81 | |
| FQ223545.1 | - | 1 - 2 | 106 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slirp
Genomic Map