Assay Details
| Gene Symbol: Noxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750205.3 | XP_003750253.1 | 3 - 4 | 654 | 74 | |
| XM_006240401.2 | XP_006240463.1 | 3 - 4 | 470 | 74 | ||
| XM_006240402.2 | XP_006240464.1 | 3 - 4 | 667 | 74 | ||
| XM_006240403.2 | XP_006240465.1 | 3 - 4 | 800 | 74 | ||
| XM_008764893.1 | XP_008763115.1 | 4 - 5 | 551 | 74 | ||
| XM_008764894.1 | XP_008763116.1 | 4 - 5 | 765 | 74 | ||
More Information
Set Membership: |
|

Noxred1
Genomic Map