Assay Details
| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191772.1 | NP_001178701.1 | 13 - 14 | 4694 | 72 | |
| XM_006240469.2 | XP_006240531.1 | 12 - 13 | 5294 | 72 | ||
| XM_006240470.2 | XP_006240532.1 | 12 - 13 | 5291 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trip11
Genomic Map