Assay Details
| Gene Symbol: Gpr68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108049.1 | NP_001101519.1 | 153 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108049.1 transcripts.
Set Membership: |
|

Gpr68
Genomic Map