Assay Details
| Gene Symbol: Otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108053.1 | NP_001101523.1 | 2 - 3 | 288 | 152 | |
| GenBank mRNA | BC168869.1 | - | 2 - 3 | 205 | 7104604 | 152 |
| FQ211938.1 | - | 2 - 3 | 274 | 152 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC168869.1,NM_001108053.1 transcripts.
Set Membership: |
|

Otub2
Genomic Map