Assay Details
| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239277.2 | XP_006239339.1 | 3 - 4 | 512 | 53 | |
| XM_578498.5 | XP_578498.3 | 3 - 4 | 579 | 53 | ||
More Information
Set Membership: |
|

Nipal3
Genomic Map