Assay Details
| Gene Symbol: Tmem200b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001068178.5 | XP_001068178.2 | 2 - 3 | 1337 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001068178.5 transcripts.
Set Membership: |
|

Tmem200b
Genomic Map