Assay Details
| Gene Symbol: Car6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134841.1 | NP_001128313.1 | 1 - 2 | 99 | 60 | |
| GenBank mRNA | DQ198382.1 | - | 1 - 2 | 99 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car6
Genomic Map