Assay Details
| Gene Symbol: LOC103689941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764128.1 | XP_008762350.1 | 4 - 5 | 890 | 90 | |
| Gene Symbol: Inpp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100755.1 | NP_001094225.1 | 9 - 10 | 1173 | 90 | |
| GenBank mRNA | BC098852.1 | - | 320 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103689941
Genomic Map