Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229520.2 | XP_006229582.1 | 10 - 11 | 855 | 119 | |
| XM_006229522.2 | XP_006229584.1 | 10 - 11 | 852 | 116 | ||
| XM_008773592.1 | XP_008771814.1 | 10 - 11 | 857 | 119 | ||
| XM_008773593.1 | XP_008771815.1 | 10 - 11 | 855 | 116 | ||
| XM_008773594.1 | XP_008771816.1 | 9 - 10 | 770 | 119 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp185
Genomic Map