Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229520.2 | XP_006229582.1 | 7 - 8 | 647 | 98 | |
| XM_006229522.2 | XP_006229584.1 | 7 - 8 | 647 | 98 | ||
| XM_008773592.1 | XP_008771814.1 | 7 - 8 | 649 | 98 | ||
| XM_008773593.1 | XP_008771815.1 | 7 - 8 | 650 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp185
Genomic Map