Assay Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004250.2 | NP_001004250.1 | 6 - 7 | 704 | 75 | |
| GenBank mRNA | BC078923.1 | - | 6 - 7 | 706 | 7106265 | 75 |
| FQ213124.1 | - | 6 - 7 | 711 | 75 | ||
| FQ215592.1 | - | 2 - 3 | 188 | 75 | ||
| FQ216746.1 | - | 6 - 7 | 707 | 75 | ||
| FQ224157.1 | - | 6 - 7 | 712 | 75 | ||
| FQ231552.1 | - | 6 - 7 | 726 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc1
Genomic Map