Assay Details
| Gene Symbol: LOC100912430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749378.3 | XP_003749426.1 | 4 - 5 | 408 | 66 | |
| Gene Symbol: Gstm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172038.1 | NP_742035.1 | 4 - 5 | 272 | 66 | |
| GenBank mRNA | U86635.1 | - | 4 - 5 | 272 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100912430
Genomic Map