Assay Details
| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106470.1 | NP_001099940.1 | 9 - 10 | 958 | 63 | |
| XM_006233266.2 | XP_006233328.1 | 9 - 10 | 1018 | 63 | ||
| GenBank mRNA | BC166789.1 | - | 9 - 10 | 864 | 7936402 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl14
Genomic Map