Assay Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191962.1 | NP_001178891.1 | 13 - 14 | 1812 | 74 | |
| XM_008761409.1 | XP_008759631.1 | 14 - 15 | 2339 | 74 | ||
| XM_008761410.1 | XP_008759632.1 | 14 - 15 | 2126 | 74 | ||
| XM_008761411.1 | XP_008759633.1 | 13 - 14 | 2035 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpsm2
Genomic Map