Assay Details
| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108950.2 | NP_001102420.2 | 8 - 9 | 959 | 131 | |
| XM_006233171.1 | XP_006233233.1 | 8 - 9 | 992 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnat2
Genomic Map