Assay Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169143.1 | NP_001162614.1 | 3 - 4 | 474 | 128 | |
| XM_006228186.2 | XP_006228248.1 | 4 - 5 | 1019 | 128 | ||
| XM_008758902.1 | XP_008757124.1 | 4 - 5 | 1039 | 128 | ||
| XM_008758903.1 | XP_008757125.1 | 3 - 4 | 524 | 128 | ||
| GenBank mRNA | FM030255.1 | - | 3 - 4 | 463 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp444
Genomic Map