Assay Details
| Gene Symbol: Qki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758777.1 | XP_008756999.1 | 3 - 4 | 917 | 114 | |
| XM_008758778.1 | XP_008757000.1 | 3 - 4 | 918 | 114 | ||
| XM_008758779.1 | XP_008757001.1 | 917 | 114 | |||
| XM_008758780.1 | XP_008757002.1 | 916 | 114 | |||
| XM_008758781.1 | XP_008757003.1 | 917 | 114 | |||
| Gene Symbol: Qk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115021.1 | NP_001108493.1 | 5 - 6 | 777 | 114 | |
| GenBank mRNA | BC158800.1 | - | 5 - 6 | 777 | 7320367 | 114 |
More Information
Updated Mapping Information
Set Membership: |
|

Qki
Genomic Map