Assay Details
| Gene Symbol: Qki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758779.1 | XP_008757001.1 | 1305 | 112 | ||
| XM_008758781.1 | XP_008757003.1 | 1281 | 112 | |||
| Gene Symbol: Qk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115021.1 | NP_001108493.1 | 7 - 8 | 1165 | 112 | |
| GenBank mRNA | BC158800.1 | - | 7 - 8 | 1165 | 7320367 | 112 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
spermatid development
axon ensheathment
positive regulation of gene expression
myelination
muscle cell differentiation
long-chain fatty acid biosynthetic process
3'-UTR-mediated mRNA destabilization
mRNA processing
regulation of translation
multicellular organism development
RNA splicing
cell differentiation
mRNA transport
RNA binding
mRNA binding
poly(A) RNA binding
SH3 domain binding
vasculogenesis
spermatid development
axon ensheathment
positive regulation of gene expression
myelination
muscle cell differentiation
long-chain fatty acid biosynthetic process
3'-UTR-mediated mRNA destabilization
mRNA processing
regulation of translation
multicellular organism development
RNA splicing
cell differentiation
mRNA transport
RNA binding
mRNA binding
poly(A) RNA binding
SH3 domain binding
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P297100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P297100 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P297100 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P297100 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P297100 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37081 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37284 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15135 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10703124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703120 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703119 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10703111 | - | - |
| Rat Genome 230 2.0 Array | 1375378_at | - | - |
| Rat Genome 230 2.0 Array | 1394495_at | - | - |
| Rat Genome 230 2.0 Array | 1375378_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394495_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375378_at | - | - |
| Affymetrix HT Rat Focus Array | 1375378_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100064701.rn.1 | TC0100008008.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375378_at | - | - |
| Rat Expression Set 230 | 1375378_PM_at | - | - |
| Rat Expression Set 230 | 1394495_PM_at | - | - |
| Rat Expression Set 230 | 1394495_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703111 | - | - |
| Rat Genome U34 Set | rc_AI102042_at | - | - |

Qki
Genomic Map