Assay Details
| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207601.1 | NP_997484.1 | 1 - 2 | 180 | 86 | |
| GenBank mRNA | BC061844.1 | - | 1 - 2 | 180 | 5621135 | 86 |
| FQ216622.1 | - | 1 - 2 | 216 | 86 | ||
| FQ220564.1 | - | 1 - 2 | 219 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Plp2
Genomic Map