Assay Details
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | HE864433.1 | - | 1846 | 65 | ||
| Gene Symbol: Ly6g5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001934.1 | NP_001001934.1 | 2 - 3 | 189 | 65 | |
| XM_008772747.1 | XP_008770969.1 | 3 - 4 | 876 | 65 | ||
| GenBank mRNA | HE864429.1 | - | 2 - 3 | 229 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk2b
Genomic Map