Assay Details
| Gene Symbol: Hspa1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212546.4 | NP_997711.1 | 1 - 2 | 150 | 73 | |
| GenBank mRNA | BC108297.1 | - | 1 - 2 | 150 | 7115041 | 73 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC108297.1,NM_212546.4 transcripts.
Set Membership: |
|

Hspa1l
Genomic Map