Assay Details
| Gene Symbol: Gtf2h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227894.1 | XP_006227956.1 | 1 - 2 | 113 | 74 | |
| GenBank mRNA | FQ212207.1 | - | 74 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_006227894.1 transcripts.
This assay is in the 5'UTR region of the XM_006227894.1 transcripts.
Set Membership: |
|

Gtf2h5
Genomic Map