Assay Details
| Gene Symbol: Il23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130410.2 | NP_569094.1 | 1 - 2 | 465 | 103 | |
| GenBank mRNA | AY055379.1 | - | 1 - 2 | 193 | 103 | |
| BC098907.1 | - | 1 - 2 | 465 | 7454618 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il23a
Genomic Map