Assay Details
| Gene Symbol: Rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765053.1 | XP_008763275.1 | 3 - 4 | 1098 | 134 | |
| XM_008765054.1 | XP_008763276.1 | 3 - 4 | 1093 | 134 | ||
| XM_008765055.1 | XP_008763277.1 | 3 - 4 | 1082 | 134 | ||
| XM_008765056.1 | XP_008763278.1 | 3 - 4 | 1098 | 134 | ||
More Information
Set Membership: |
|

Rdh5
Genomic Map