Assay Details
| Gene Symbol: Rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765056.1 | XP_008763278.1 | 4 - 5 | 1256 | 62 | |
| GenBank mRNA | FQ218828.1 | - | 4 - 5 | 823 | 62 | |
More Information
Set Membership: |
|

Rdh5
Genomic Map