Assay Details
| Gene Symbol: Pacrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077677.1 | NP_001071145.1 | 4 - 5 | 814 | 107 | |
| GenBank mRNA | BC127534.1 | - | 4 - 5 | 814 | 7133203 | 107 |
More Information
Set Membership: |
|

Pacrg
Genomic Map