Assay Details
| Gene Symbol: Dennd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126289.1 | NP_001119761.1 | 11 - 12 | 1428 | 56 | |
| GenBank mRNA | BC160823.1 | - | 11 - 12 | 1428 | 7380256 | 56 |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd1c
Genomic Map