Assay Details
| Gene Symbol: Pot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001073777.4 | XP_001073777.2 | 12 - 13 | 1662 | 164 | |
| XM_006244268.2 | XP_006244330.1 | 13 - 14 | 1728 | 164 | ||
| XM_006244269.2 | XP_006244331.1 | 13 - 14 | 1957 | 164 | ||
| XM_006244270.2 | XP_006244332.1 | 13 - 14 | 2242 | 164 | ||
| XM_006244271.2 | XP_006244333.1 | 13 - 14 | 2254 | 164 | ||
| XM_008766793.1 | XP_008765015.1 | 12 - 13 | 2087 | 164 | ||
| XM_008766794.1 | XP_008765016.1 | 9 - 10 | 2346 | 164 | ||
More Information
Set Membership: |
|

Pot1b
Genomic Map