Assay Details
| Gene Symbol: Pot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244268.2 | XP_006244330.1 | 3 - 4 | 229 | 139 | |
| XM_006244269.2 | XP_006244331.1 | 3 - 4 | 458 | 139 | ||
| XM_006244270.2 | XP_006244332.1 | 3 - 4 | 743 | 139 | ||
| XM_006244271.2 | XP_006244333.1 | 3 - 4 | 755 | 139 | ||
| XM_008766793.1 | XP_008765015.1 | 3 - 4 | 744 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006244268.2, XM_006244269.2, XM_006244270.2, XM_006244271.2, XM_008766793.1 transcripts.
Set Membership: |
|

Pot1b
Genomic Map